Anti-PDE4B Goat Polyclonal Antibody

Supplier: ProSci
42-925
10115-106EA 701.76 USD
10115-106
Anti-PDE4B Goat Polyclonal Antibody
Antibodies
Western Blot: Approx 98 kDa band observed in Human Uterus lysates (calculated MW of 83.3 kDa according to NP_002591.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Mehats et al, Endocrinology. 1999 Jul;140(7):3228-37. PMID: 10385419).Recommended concentration: 1-3 ?g/mL.

PDE4B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PDE4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR