Anti-SLC5A1 Goat Polyclonal Antibody

Supplier: ProSci
42-919
10115-098EA 701.94 USD
10115-098
Anti-SLC5A1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 4000. Western Blot: Preliminary experiments in rat kidney, duodenum, Jejunum and colon lysates gave no specific signal but low background (at antibody concentration up to 1 ?g/mL).

Slc5a1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Slc5a1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR