Anti-HP Goat Polyclonal Antibody

Supplier: ProSci
42-911
10112-324EA 701.76 USD
10112-324
Anti-HP Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 8000. Western Blot: Approximately 37+40 kDa bands observed in human liver lysates (calculated MW of 45.2 kDa according to NP_005134.1 and 38.5 kDa according to NP_001119574.1). Recommended concentration: 0.1-0.3 ?g/mL.

HP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR