Anti-APOH Goat Polyclonal Antibody

Supplier: ProSci
42-910
10115-088EA 701.94 USD
10115-088
Anti-APOH Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1000. Western Blot: Approximately 50 kDa band observed in human liver, kidney and placenta lysates (calculated MW of 38.3 kDa according to NP_000033.2). Recommended concentration: 0.2-0.6 ?g/mL.

APOH antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: APOH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR