Anti-NPR3 Goat Polyclonal Antibody

Supplier: ProSci
42-909
10115-086EA 701.76 USD
10115-086
Anti-NPR3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 16000. Western Blot: Approximately 52 kDa band observed in human placenta lysates (calculated MW of 59.8 kDa according to NP_000899.1). Recommended concentration: 1-3 ?g/mL.

NPR3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NPR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR