Anti-ULK3 Goat Polyclonal Antibody

Supplier: ProSci
42-905
10112-320EA 701.94 USD
10112-320
Anti-ULK3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 32000. Western Blot: Approximately 75 kDa band in human brain, heart and testis lysates after 0.1 ?g/mL antibody staining.

ULK3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ULK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR