Anti-cyp1a1 Goat Polyclonal Antibody

Supplier: ProSci
42-902
10115-076EA 701.94 USD
10115-076
Anti-cyp1a1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 64000. Western Blot: Approximately 55 kDa band observed in rat lung lysates (calculated MW of 59.2 kDa according to mouse NP_001129531.1 and 59.4 kDa according to rat NP_036672.). Recommended concentration: 0.1-0.3 ?g/mL.

cyp1a1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: cyp1a1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR