Anti-Cyp1a2 Goat Polyclonal Antibody

Supplier: ProSci
42-901
10115-074EA 701.94 USD
10115-074
Anti-Cyp1a2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 32000. Western Blot: Approximately 60 kDa band observed in mouse fetal liver lysates (calculated MW of 58.2 kDa according to NP_034123.1). Recommended concentration: 1-3 ?g/mL.

Cyp1a2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Cyp1a2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR