Anti-POU3F1 Goat Polyclonal Antibody

Supplier: ProSci
42-897
10112-316EA 701.94 USD
10112-316
Anti-POU3F1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 8000. Western Blot: Approximately 48 kDa band observed in A549 lysate (calculated MW of 45.5 kDa according to NP_002690.3). Recommended concentration: 1-3 ?g/mL.

POU3F1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: POU3F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR