Anti-SYNGAP Goat Polyclonal Antibody

Supplier: ProSci
42-895
10112-312EA 701.94 USD
10112-312
Anti-SYNGAP Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 32000. Western Blot: Approximately 150 kDa band observed in mouse brain lysates (calculated MW of 148 kDa according to human NP_006763.2 and mouse XP_990642.3). Recommended concentration: 0.03-0.1 ?g/mL.

SYNGAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SYNGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR