Anti-ZCCHC11 Goat Polyclonal Antibody

Supplier: ProSci
42-885
10112-306EA 701.94 USD
10112-306
Anti-ZCCHC11 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 32000. Western Blot: Approximately 170 kDa band observed in rat brain lysates (calculated MW of 163 kDa according to rat NP_001101423.1 and 185 kDa according to mouse NP_780681.2). Recommended concentration: 0.03-0.1 ?g/mL.

Zcchc11 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Zcchc11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR