Anti-TGM4 Goat Polyclonal Antibody

Supplier: ProSci

42-884
10115-054EA 560.36 USD
10115-054
Anti-TGM4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 16000. Western Blot: Approximately 75 kDa band observed in HeLa lysate (calculated MW of 77.1 kDa according to NP_003232.2). Recommended concentration: 1-3 ?g/mL.

TGM4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TGM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR