Anti-CRLS1 Goat Polyclonal Antibody

Supplier: ProSci
42-880
10115-048EA 701.94 USD
10115-048
Anti-CRLS1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 64000. Western Blot: Approximately 30 kDa band observed in HEK293 lysate (calculated MW of 32.6 kDa according to NP_061968.1). Recommended concentration: 0.2-0.6 ?g/mL.

CRLS1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CRLS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR