Anti-Cyp2d5 Goat Polyclonal Antibody

Supplier: ProSci
42-876
10115-044EA 701.94 USD
10115-044
Anti-Cyp2d5 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 32000. Western Blot: Approximately 65 kDa band observed in rat spleen lysates (calculated MW of 57.1 kDa according to NP_775426.1). Recommended concentration: 1-3 ?g/mL.

Cyp2d5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Cyp2d5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR