Anti-SCN9A Goat Polyclonal Antibody

Supplier: ProSci
42-870
10115-034EA 552.17 USD
10115-034
Anti-SCN9A Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 80 kDa band in human cerebellum, mouse brain and rat ovary lysates after 0.1 ?g/mL antibody staining. The 80 kDa band was successfully blocked by incubation with the immunizing peptide.

SCN9A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SCN9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR