Anti-GAD1 Goat Polyclonal Antibody

Supplier: ProSci
42-864
10115-026EA 701.94 USD
10115-026
Anti-GAD1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 70 kDa band observed in mouse brain lysates (calculated MW of 66.6 kDa according to mouse NP_032103.2). Recommended concentration: 1-3 ?g/mL.

GAD1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR