Anti-FGF23 Goat Polyclonal Antibody

Supplier: ProSci
42-859
10112-288EA 701.94 USD
10112-288
Anti-FGF23 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 27+38 kDa bands observed in mouse brain lysates (calculated MW of 27.7 kDa according to NP_073148.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Garringer et al, Am J Physiol Endocrinol Metab. 2008 Oct;295 (4) :E929-37. PMID: 18682534). Recommended concentration: 0.5-2 ?g/mL.

Fgf23 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Fgf23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR