Anti-VIPR1 Goat Polyclonal Antibody

Supplier: ProSci
42-856
10115-018EA 701.94 USD
10115-018
Anti-VIPR1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 20 kDa band in mouse small intestine lysates after 0.1 ?g/mL antibody staining.1 kDa according to NP_035833.2. The 20 kDa band was successfully blocked by incubation with the immunizing peptide.

Vipr1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Vipr1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR