Anti-HTR3B Goat Polyclonal Antibody

Supplier: ProSci
42-854
10112-284EA 701.94 USD
10112-284
Anti-HTR3B Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 50 kDa band observed in rat brain lysates (calculated MW of 50.3 kDa according to NP_071525.1). Recommended concentration: 0.03-0.1 ?g/mL.

Htr3b antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Htr3b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR