Anti-APOBEC3D Goat Polyclonal Antibody

Supplier: ProSci
42-853
10115-016EA 701.94 USD
10115-016
Anti-APOBEC3D Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 45 kDa band observed in A549 lysate (calculated MW of 46.6 kDa according to NP_689639.2). Recommended concentration: 0.3-1 ?g/mL.

APOBEC3D antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: APOBEC3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR