Anti-MKRN1 Goat Polyclonal Antibody

Supplier: ProSci

42-850
10115-010EA 560.36 USD
10115-010
Anti-MKRN1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: In transfected HEK293 transiently expressing full-length human MKRN1 (myc and DYKDDDDK tagged), a band of approximately 60 kDa was observed. No bands were observed in mock-transfected HEK293 and the same band was observed using anti-DYKDDDDK tag antibody. Recommended concentration, 0.3- 1 ?g/mL.

MKRN1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MKRN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR