Anti-ERO1A Goat Polyclonal Antibody

Supplier: ProSci
42-847
10115-004EA 701.94 USD
10115-004
Anti-ERO1A Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 70 kDa band observed in lysates of cell lines A431, HeLa, Jurkat (calculated MW of 54.4 kDa according to NP_055399.1). The observed molecular weight corresponds to the glycosylated form. Recommended concentration: 0.1-0.3 ?g/mL.

ERO1L antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ERO1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR