Anti-SAP30L Goat Polyclonal Antibody

Supplier: ProSci
42-835
10114-986EA 701.94 USD
10114-986
Anti-SAP30L Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 18 kDa band observed in rat spleen lysates (calculated MW of 20.8 kDa according to rat XP_340804.4). Recommended concentration: 0.3-1 ?g/mL.

Sap30L antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Sap30L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR