Anti-PRKAA2 Goat Polyclonal Antibody

Supplier: ProSci
42-828
10114-974EA 701.94 USD
10114-974
Anti-PRKAA2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 65 kDa band observed in human heart and liver lysates (calculated MW of 62.3 kDa according to NP_006243.2). Recommended concentration: 1-3 ?g/mL.

PRKAA2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PRKAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR