Anti-MYO5A Goat Polyclonal Antibody

Supplier: ProSci
42-827
10114-972EA 701.94 USD
10114-972
Anti-MYO5A Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 200 kDa band observed in Jurkat lysate (calculated MW of 215 kDa according to NP_000250.3). Recommended concentration: 1-3 ?g/mL. Immunofluorescence: Transverse sections of mouse skeletal muscle shows staining of the fibre membrane and neuromuscular junction. The observed pattern corresponds to earlier findings in literature with different antibodies (Röder et al, Proc Natl Acad Sci U S A. 2010 Feb 2;107 (5) :2031-6; PMID: 20133847). Recommended concentration: 1-3 3 ?g/mL.

MYO5A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MYO5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR