Anti-SIGLEC6 Goat Polyclonal Antibody

Supplier: ProSci
42-822
10114-964EA 701.94 USD
10114-964
Anti-SIGLEC6 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Experiments gave bands at approximately 60 kDa and 35 kDa in human placenta lysates. These bands correspond to earlier findings with different antibodies from other commercial sources. This protein has a calculated MW of 49.9 kDa according to NP_001236.4. Recommended concentration: 0.5-1.5 ?g/mL.

SIGLEC6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SIGLEC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR