Anti-NDUFS8 Goat Polyclonal Antibody

Supplier: ProSci
42-813
10114-950EA 701.94 USD
10114-950
Anti-NDUFS8 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4000. Western Blot: Approximately 25 kDa band observed in mouse and rat heart lysates (calculated MW of 24.0 kDa according to mouse NP_659119.2 and rat NP_001099792.1). Recommended concentration: 0.1-0.3 ?g/mL.

NDUFS8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NDUFS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR