Anti-HCN3 Goat Polyclonal Antibody

Supplier: ProSci
42-810
10114-944EA 701.94 USD
10114-944
Anti-HCN3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 80 kDa band observed in mouse brain lysates (calculated MW of 86.6 kDa according to mouse NP_032253.1). Recommended concentration: 0.01-0.03 ?g/mL.

HCN3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HCN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR