Anti-PEX1 Goat Polyclonal Antibody

Supplier: ProSci
42-803
10114-934EA 701.94 USD
10114-934
Anti-PEX1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Preliminary experiments in human liver and kidney lysates gave no specific signal but low background (at antibody concentration up to 1 ?g/mL).

Pex1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Pex1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR