Anti-FANCE Goat Polyclonal Antibody

Supplier: ProSci
42-799
10114-930EA 701.76 USD
10114-930
Anti-FANCE Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 70 kDa band in human adipose and uterus lysates after 0.1 ?g/mL antibody staining.7 kDa according to NP_068741.1. The 70 kDa band was successfully blocked by incubation with the immunizing peptide.

FANCE antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FANCE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR