Anti-HOXB9 Goat Polyclonal Antibody

Supplier: ProSci
42-797
10114-926EA 701.94 USD
10114-926
Anti-HOXB9 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4000. Western Blot: Approximately 27 kDa band observed in human cerebellum and mouse spinal cord lysates (calculated MW of 28.1 kDa according to human NP_076922.1 and 28.0 kDa according to mouse NP_032296.2). Recommended concentration: 0.3-1 ?g/mL.

HOXB9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HOXB9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR