Anti-HOXA1 Goat Polyclonal Antibody

Supplier: ProSci
42-791
10114-918EA 701.94 USD
10114-918
Anti-HOXA1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. In transfected HEK293 transiently expressing human HOXA1 (MYC and DYKDDDDK tagged), a band of Western Blot: Approximately 19 kDa is observed. No bands are observed in mock-transfected HEK293 and the same band is observed using anti-MYC Tag antibody. Recommended concentration, 1-3 ?g/mL.

HOXA1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HOXA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR