Anti-CANX Goat Polyclonal Antibody

Supplier: ProSci
42-774
10114-886EA 701.94 USD
10114-886
Anti-CANX Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 100 kDa band observed in mouse heart, human cerebellum, NIH3T3 and HeLa lysates (calculated MW of 67.6 kDa according to human NP_001737.1 and 67.2 kDa according to mouse NP_001103969.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Oliver et al., J Biol Chem. 1996 Jun 7;271 (23) :13691-6., PMID: 8662691). Recommended concentration: 0.3-1 ?g/mL. Immunohistochemistry: In paraffin embedded human kidney shows textured staining in the cytoiplasm of DCT cells,. Recommended concentration, 3-6 ?g/mL.

CANX antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CANX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR