Anti-P2RX7 Goat Polyclonal Antibody

Supplier: ProSci
42-772
10114-882EA 701.94 USD
10114-882
Anti-P2RX7 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 37 kDa and 70 kDa band observed in human frontal cortex lysates (calculated MW of 68.6 kDa according to NP_002553.2). Recommended concentration: 1-3 ?g/mL. Immunohistochemisry: In paraffin embedded human brain cortex shows membranous staining of cell bodies and processes. Recommended concentration, 3-6 3 ?g/mL.

P2RX7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: P2RX7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR