Anti-ACO1 Goat Polyclonal Antibody

Supplier: ProSci
42-768
10114-876EA 701.94 USD
10114-876
Anti-ACO1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 100 kDa band observed in human liver lysates (calculated MW of 98.4 kDa according to NP_002188.1). Recommended concentration: 0.1-0.3 ?g/mL.

ACO1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ACO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR