Anti-SATB1 Goat Polyclonal Antibody

Supplier: ProSci
42-764
10114-868EA 701.94 USD
10114-868
Anti-SATB1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Experiments gave bands at approximately 110 kDa and 85 kDa in Molt4 lysate after 0.3 ?g/mL antibody staining. The observed 110 kDa band corresponds to earlier findings in literature with different antibodies (De Belle et al, J Cell Biol. 1998 Apr 20;141 (2) :335-48.; PMID: 9548713). This protein has a calculated MW of 86.0 kDa according to NP_002962.1. Recommended concentration: 0.3-1 ?g/mL.

SATB1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SATB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR