Anti-CHRNA4 Goat Polyclonal Antibody

Supplier: ProSci
42-755
10113-678EA 701.94 USD
10113-678
Anti-CHRNA4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 70 kDa band observed in rat brain lysates (calculated MW of 70.0 kDa according to human NP_000735.1and of 70.2 kDa according to rat NP_077330.1). Recommended concentration: 1-3 ?g/mL.
an additional band of unknown identity was also consistently observed at 22 kDa . This band was successfully blocked by incubation with the immunizing peptide.

CHRNA4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CHRNA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR