Anti-TRPV2 Goat Polyclonal Antibody

Supplier: ProSci
42-751
10113-672EA 701.94 USD
10113-672
Anti-TRPV2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 100 kDa band observed in rat brain lysates (calculated MW of 86.0 kDa according to mouse NP_035836.2 and of 86.7 kDa acording to rat NP_058903.2). Recommended concentration: 1-3 ?g/mL.

TRPV2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TRPV2*
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR