Anti-ABCC8 Goat Polyclonal Antibody

Supplier: ProSci
42-749
10114-850EA 701.94 USD
10114-850
Anti-ABCC8 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 170 kDa band observed in human cerebellum lysates (calculated MW of 177 kDa according to NP_000343.2). Recommended concentration: 0.5-1.5 ?g/mL. an additional band of unknown identity was also consistently observed at 23 kDa . This band was successfully blocked by incubation with the immunizing peptide.

ABCC8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ABCC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR