Anti-FGFR2 Goat Polyclonal Antibody

Supplier: ProSci
42-748
10114-848EA 625.43 USD
10114-848
Anti-FGFR2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 100 kDa band observed in human a549 lysate (calculated MW of 92.0 kDa according to NP_000132.1 and of 92.1 kDa according to NP_075259.2). Recommended concentration: 0.3-1 ?g/mL.

FGFR2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FGFR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR