Anti-DACH1 Goat Polyclonal Antibody

Supplier: ProSci

42-747
10114-846EA 560.36 USD
10114-846
Anti-DACH1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 75 and 52 kDa bands observed in human kidney lysates (calculated MW of 73.0 kDa according to NP_542937.1 and of 51.9 kDa according to NP_004383.2). Recommended concentration: 0.1-0.3 ?g/mL.

DACH1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DACH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR