Anti-APOBEC3C Goat Polyclonal Antibody

Supplier: ProSci
42-743
10114-838EA 701.94 USD
10114-838
Anti-APOBEC3C Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1000. Western Blot: Approximately 22 kDa band observed in lysates of Daudi lysate (calculated MW of 22.8 kDa according to NP_055323.2). Recommended concentration: 0.5-1.5 ?g/mL.

APOBEC3C antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: APOBEC3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR