Anti-NTN1 Goat Polyclonal Antibody

Supplier: ProSci
42-733
10114-822EA 701.94 USD
10114-822
Anti-NTN1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 70 kDa band observed in human cerebellum, mouse and rat brain lysates (calculated MW of 67.7 kDa according to human NP_004813.1 and 67.8 kDa according to rat NP_446183.1). Recommended concentration: 0.5-1.5 ?g/mL.

NTN1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR