Anti-PRPF31 Goat Polyclonal Antibody

Supplier: ProSci
42-726
10113-662EA 625.43 USD
10113-662
Anti-PRPF31 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 60 kDa band observed in lysates of cell line A431 (calculated MW of 55.4 kDa according to NP_056444.3. Recommended concentration: 0.3-1.0 ?g/mL.

PRPF31 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PRPF31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR