Anti-ILK Goat Polyclonal Antibody

Supplier: ProSci
42-724
10114-810EA 701.94 USD
10114-810
Anti-ILK Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 52 kDa band observed in mouse kidney lysates and in lysates of cell line NIH3T3 (calculated MW of 51.4 kDa according to human NP_004508.1 and mouse NP_034692.2). Recommended concentration: 0.5-2 ?g/mL.

ILK antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ILK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR