Anti-DENND4C Goat Polyclonal Antibody

Supplier: ProSci

42-719
10114-804EA 560.36 USD
10114-804
Anti-DENND4C Goat Polyclonal Antibody
Antibodies
Approximately 210 kDa band observed in mouse spleen lysates (calculated MW of 211 kDa according to NP_908976.1). Recommended concentration: 0.5-2 ?g/mL.

Dennd4c antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Dennd4c
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR