Anti-NOX5 Goat Polyclonal Antibody

Supplier: ProSci
42-714
10114-798EA 701.94 USD
10114-798
Anti-NOX5 Goat Polyclonal Antibody
Antibodies
Approximately 90 kDa band observed in human testis lysates (calculated MW of 86.4 kDa according to NP_078781.3). Recommended concentration: 0.5-2 ?g/mL.

NOX5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NOX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR