Anti-SYT2 Goat Polyclonal Antibody

Supplier: ProSci

42-713
10114-796EA 560.36 USD
10114-796
Anti-SYT2 Goat Polyclonal Antibody
Antibodies
Approximately 50 kDa band observed in mouse lung lysates (calculated MW of 47.2 kDa according to mouse NP_033333.2). Recommended concentration: 0.1-0.3 ?g/mL.

SYT2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SYT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR