Anti-NPAS4 Goat Polyclonal Antibody

Supplier: ProSci
42-711
10114-792EA 701.94 USD
10114-792
Anti-NPAS4 Goat Polyclonal Antibody
Antibodies
Approximately 90 kDa band observed in human frontal cortex lysates (calculated MW of 87.1 kDa according to NP_849195.2). Recommended concentration: 0.3-1 ?g/mL.

NPAS4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NPAS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR