Anti-GPM6A Goat Polyclonal Antibody

Supplier: ProSci
42-708
10114-786EA 701.94 USD
10114-786
Anti-GPM6A Goat Polyclonal Antibody
Antibodies
Approximately 30 kDa band observed in rat brain lysates. an approximately 80 kDa band was observed in human, mouse and rat brain lysates (calculated MW of 31.2 kDa according to NP_005268.1). The observed 80 kDa band corresponds to the glycosylated form. Recommended concentration: 0.5-2 ?g/mL.

GPM6A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPM6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR